missing translation for 'onlineSavingsMsg'
Learn More

Salicylic Acid, 99.9+%, Certified AR for Analysis, Fisher Chemical

Product Code. 11413443 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11413443 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11413443 Supplier Fisher Chemical Supplier No. S/0282/53
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 69-72-7
Melting Point 158°C
Color White
pH 3.6
Boiling Point 211°C
Packaging Plastic powder jar
Molecular Formula C7H6O3
MDL Number MFCD00002439
Quantity 500 g
Synonym salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC=C1O
IUPAC Name 2-hydroxybenzoic acid
Molecular Weight (g/mol) 138.12
PubChem CID 338
ChEBI CHEBI:16914
Formula Weight 138.12g/mol
Vapor Pressure 0.3mbar 95
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Salicylic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.