missing translation for 'onlineSavingsMsg'
Learn More

Salicylsalicylic acid, 98%

Product Code. 13448358 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13448358 100 g Glass bottle 100g
10336590 25 g Glass bottle 25g
13438358 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13448358 Supplier Thermo Scientific Acros Supplier No. 133401000
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 552-94-3 | C14H10O5 | 258.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-94-3
Molecular Formula C14H10O5
Molecular Weight (g/mol) 258.22
MDL Number MFCD00020252
InChI Key WVYADZUPLLSGPU-UHFFFAOYSA-N
Synonym salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal
PubChem CID 5161
ChEBI CHEBI:9014
IUPAC Name 2-(2-hydroxybenzoyl)oxybenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O

Specifications

Melting Point 139.0°C to 151.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 10,II,54
Merck Index 15,8477
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in alcohol and ether, sparingly soluble in benzene, slightly soluble in acetone
Formula Weight 258.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Salicylsalicylic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Salicylsalicylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.