missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salivary Amylase Alpha Antibody (1G4D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61869-0.025ml
This item is not returnable.
View return policy
Description
Salivary Amylase Alpha Monoclonal specifically detects Salivary Amylase Alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Salivary Amylase Alpha | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| 1,4-alpha-D-glucan glucanohydrolase 1, alpha-amylase 1, AMY1, AMY1B, AMY1C, amylase, alpha 1A (salivary), amylase, alpha 1A; salivary, amylase, salivary, alpha-1A, EC 3.2.1.1, glycogenase, Salivary alpha-amylase, salivary amylase alpha 1A | |
| Mouse | |
| 57.8 kDa | |
| 0.025 mL | |
| 276 | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 1G4D8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| AMY1A | |
| Purified recombinant fragment of human AMY1A (AA: 172-284) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction