missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SALL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
234.00 € - 326.00 €
Specifications
| Antigen | SALL4 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:300-1:10000 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643482
|
Novus Biologicals
NBP2-98704-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606382
|
Novus Biologicals
NBP2-98704-50ul |
50 μg |
234.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SALL4 Polyclonal antibody specifically detects SALL4 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| SALL4 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| dJ1112F19.1, DRRS, HSAL4, MGC133050, sal-like 4 (Drosophila), sal-like protein 4, Zinc finger protein 797, Zinc finger protein SALL4, ZNF797sal (Drosophila)-like 4 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SALL4. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 57167 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title