missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Salmeterol xinafoate

Product Code. 15793457 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15793457 100 mg 100mg
15703467 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15793457 Supplier Thermo Scientific Chemicals Supplier No. 462171000
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94749-08-3
Molecular Formula C36H45NO7
Molecular Weight (g/mol) 603.76
MDL Number MFCD00897708
InChI Key XTZNCVSCVHTPAI-UHFFFAOYNA-N
Synonym salmeterol xinafoate, salmetedur, arial, serevent diskus, asmerole, betamican, dilamax, inaspir, ultrabeta, beglan
PubChem CID 56801
IUPAC Name 2-(hydroxymethyl)-4-[1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]phenol;1-hydroxynaphthalene-2-carboxylic acid
SMILES OC(=O)C1=CC=C2C=CC=CC2=C1O.OCC1=CC(=CC=C1O)C(O)CNCCCCCCOCCCCC1=CC=CC=C1

Specifications

Melting Point 126.0°C to 128.0°C
Quantity 100 mg
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dmso, methanol
Formula Weight 603.75
Chemical Name or Material Salmeterol xinafoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.