missing translation for 'onlineSavingsMsg'
Learn More

Salmeterol xinafoate, MedChemExpress

Product Code. 16493367
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493367 50 mg 50mg
16487348 10 mg 10mg
16457316 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493367 Supplier Medchem Express Supplier No. HY17453
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Salmeterol (GR 33343X) xinafoate is a potent and selective human β2 adrenoceptor agonist. Salmeterol shows potent stimulation of cAMP accumulation in CHO cells expressing human β2, β1 and β3 adrenoceptors with pEC50s of 9.6, 6.1, and 5.9, respectively.

Salmeterol (GR 33343X) xinafoate is a potent and selective human β2 adrenoceptor agonist. Salmeterol shows potent stimulation of cAMP accumulation in CHO cells expressing human β2, β1 and β3 adrenoceptors with pEC50s of 9.6, 6.1, and 5.9, respectively.

Specifications

Chemical Name or Material Salmeterol xinafoate
CAS 94749-08-3
Color White
Purity Grade Notes Research
Molecular Formula C36H45NO7
Quantity 50 mg
Synonym GR 33343X xinafoate
Solubility Information DMSO : ≥ 50 mg/mL (82.82 mM)
SMILES O=C(C1=CC=C2C=CC=CC2=C1O)O.OC3=C(CO)C=C(C(CNCCCCCCOCCCCC4=CC=CC=C4)O)C=C3
Molecular Weight (g/mol) 603.75
Formula Weight 603.75
Percent Purity 99.83%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.