missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Salmonella Chromogenic Medium

Product Code. 12977178
This item is not returnable. View return policy
Product Code. 12977178 Supplier Thermo Scientific™ Supplier No. PO0958A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and differentiate Salmonella species from other Enterobacteriaceae with this selective and differential ready-to-use medium.

Isolate and differentiate Salmonella species from other Enterobacteriaceae with ready-to-use Thermo Scientific™ Oxoid™ Salmonella Chromogenic Medium. Designed to identify Salmonella species based on their utilization of one chromogenic substrate. The addition of bile salts, novobiocin and cefsulodin inhibit Gram-negative organisms, Proteus and Pseudomonad growth respectively.

Isolate and differentiate Salmonella species from other Enterobacteriaceae with Salmonella Chromogenic Medium.

  • Easy to read combination of two chromogens visually differentiates Salmonella from other Enterobacteriaceae.
  • Selective for Salmonella due to the additions of bile salts, novobiocin and cefsulodin.
  • Ready-to-use convenience of prepared media.

Traditionally, media used to differentiate Salmonella species from other members of the family Enterobacteriaceae depend upon the ability of Salmonella species to produce hydrogen sulphide coupled with their ability to ferment lactose.1, 2 These are, however, essentially inadequate methods with a significant number of species not exhibiting these characteristics. Chromogenic media have been developed for rapid and more reliable identification of Salmonella.

Salmonella Chromogenic Medium combines two chromogens for the detection of Salmonella species, 5-Bromo-6-Chloro-3-Indolyl caprylate (Magenta-caprylate) and 5-Bromo-4-Chloro-3-Indolyl b-D galactopyranoside (X-gal). X-gal is a substrate for the enzyme b-D-galactosidase. Most Enterobacteriaceae are b-D-galactosidase positive and produce blue colored colonies on Salmonella Chromogenic Medium. Hydrolysis of the chromogen Mag-caprylate by lactose negative Salmonella species results in magenta colonies which are easily distinguished from the blue colored colonies of other organisms.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2°C to 10°C
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 18g ± 1.0g
Quantity 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.