Learn More
Invitrogen™ Salmonella LPS Monoclonal Antibody (A99H)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518258
Description
This antibody is specific for common LPS core of all Salmonellae O-serogroups tested: A, B, C1, C2, D, E1, E3, E4, F, G1 and G2. This does not cross-react with E. coli (mix), Klebsiella, Citrobacter, Pseudomonas, Yersinia, Shigella, Proteus or Legionella. Purity is >90%. Recommended pairs for ELISA: MA5-18258 (capture) and MA5-18259 or MA5-18260 (conjugated, detection).
The genus Salmonella is a member of the family Enterobacteriaceae. The genus is composed of Gram negative bacilli that are facultative and flagellated (motile). Salmonellae possess 3 major surface antigens; the H or flagellar antigen (phase 1 and 2), the O or somatic antigen (part of the LPS moiety) and the Vi or capsular antigen (referred to as K in other Enterobacteriaceae). Salmonellae also possess the LPS endotoxin characteristic of Gram negative bacteria. This LPS is composed of an O polysaccharide (O antigen) an R core and the endotoxic inner Lipid A. Endotoxins evoke fever and can activate complement, kinin and clotting factors.
Specifications
| Salmonella LPS | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2a |
| ELISA, Immunocytochemistry | |
| A99H | |
| Unconjugated | |
| Salmonellae | |
| Salmonellae sp. native protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.