missing translation for 'onlineSavingsMsg'
Learn More

Salsalate, MedChemExpress

Product Code. 16418706
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418706 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418706 Supplier Medchem Express Supplier No. HYB1245
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Salsalate, a non-acetylated salicylate, is an effective antirheumatic drug that bypasses gastric absorption and also avoids cyclooxygenase inhibition. Salsalate has anti-inflammatory activity and reduces glucose levels, insulin resistance, and cytokine expression. Salsalate can be used in the research of type 2 diabetes.

Salsalate, a non-acetylated salicylate, is an effective antirheumatic drug that bypasses gastric absorption and also avoids cyclooxygenase inhibition. Salsalate has anti-inflammatory activity and reduces glucose levels, insulin resistance, and cytokine expression. Salsalate can be used in the research of type 2 diabetes.

Specifications

Chemical Name or Material Salsalate
CAS 552-94-3
Color White
Purity Grade Notes Research
Molecular Formula C14H10O5
Quantity 100 mg
Synonym Salicylsalicylic acid Disalicylic acid
Solubility Information DMSO : ≥ 100 mg/mL (387.25 mM) ∣H2O : 0.67 mg/mL (2.59 mM; Need ultrasonic)
SMILES O=C(OC1=CC=CC=C1C(O)=O)C2=CC=CC=C2O
Molecular Weight (g/mol) 258.23
Formula Weight 258.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.