Learn More
Novus Biologicals™ SAMD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30871-25ug
 View more versions of this product
Product Code. 30066992
  • 213.00 € / 25µg
Description
SAMD8 Polyclonal antibody specifically detects SAMD8 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SAMD8
Polyclonal
Western Blot : 5-10 μg/mL, ELISA
FLJ25082, SAM domain-containing protein 8, SMSr, sphingomyelin synthase related, sphingomyelin synthase-related protein 1, sterile alpha motif domain containing 8, Sterile alpha motif domain-containing protein 8
Full-length recombinant SAMD8 protein
25 μg
Lipid and Metabolism
142891
Store at -20°C. Avoid freeze/thaw cycles.
ELISA, Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
Primary
Human
Purified