Learn More
Invitrogen™ SAMM50 Recombinant Rabbit Monoclonal Antibody (24GB1135)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557702
 View more versions of this product
Product Code. 30274975
  • 564.00 € / 100µL
Description
SAMM50 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
SAMM50 is a component of the sorting and assembly machinery (SAM) complex of the outer mitochondrial membrane. The SAM complex has a role in integrating beta-barrel proteins into the outer mitochondrial membrane (Humphries et al, 2005).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SAMM50
Recombinant Monoclonal
2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q6AXV4, Q8BGH2, Q9Y512
Samm50
A synthesized peptide derived from human SAMM50 (aa 350-390).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1135
Unconjugated
Samm50
1110030L07Rik; CGI-51; OMP85; SAM50; SAMM50; SAMM50 sorting and assembly machinery component; sorting and assembly machinery 50kDa; sorting and assembly machinery component 50 homolog; sorting and assembly machinery component 50 homolog (S. cerevisiae); sorting and assembly machinery component 50 homolog A; TOB55; Transformation-related gene 3 protein; TRG3; TRG-3; YNL026W
Rabbit
Affinity chromatography
RUO
25813, 300111, 68653
-20°C