missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Samuraciclib hydrochloride, MedChemExpress
Description
Samuraciclib hydrochloride displays 45-, 15-, 230- and 30-fold selectivity over CDK1, CDK2 (IC50 of 578 nM), CDK5 and CDK9, respectively. Samuraciclib hydrochloride inhibits the growth of breast cancer cell lines with GI50 values between 0.2-0.3 μM. Samuraciclib hydrochloride has anti-tumor effects.
Specifications
Specifications
| Chemical Name or Material | Samuraciclib hydrochloride |
| Physical Form | Powder |
| CAS | 1805789-54-1 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H31ClN6O |
| For Use With (Application) | Cancer-Kinase/protease |
| Synonym | CT7001 hydrochlorideICEC0942 hydrochloride |
| Solubility Information | H2O : 55 mg/mL (127.62 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (116.02 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?