missing translation for 'onlineSavingsMsg'
Learn More

SANT-1, MedChemExpress

Product Code. 16465597
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465597 5 mg 5mg
16442858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16465597 Supplier Medchem Express Supplier No. HY100224
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SANT-1, a potent Smo antagonist, inhibits Hedgehog signaling. SANT-1 shows IC50s of 20 nM and 30 nM in Shh-LIGHT2 and SmoA1-LIGHT2 assay, respectively.

SANT-1, a potent Smo antagonist, inhibits Hedgehog signaling. SANT-1 shows IC50s of 20 nM and 30 nM in Shh-LIGHT2 and SmoA1-LIGHT2 assay, respectively.

Specifications

Chemical Name or Material SANT-1
Physical Form Powder
CAS 304909-07-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H27N5
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 62.5 mg/mL (167.34 mM; Need ultrasonic)
SMILES CC1=C(/C=N/N2CCN(CC3=CC=CC=C3)CC2)C(C)=NN1C4=CC=CC=C4
Molecular Weight (g/mol) 373.49
Formula Weight 373.49
Percent Purity 99.71%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.