missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAP155 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47291
This item is not returnable.
View return policy
Description
SAP155 Polyclonal specifically detects SAP155 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| SAP155 | |
| Polyclonal | |
| Western Blot 1:100 - 1:250, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Hsh155, pre-mRNA processing 10, pre-mRNA splicing factor SF3b, 155 kDa subunit, Pre-mRNA-splicing factor SF3b 155 kDa subunit, Prp10, PRPF10, SAP 155, SAP155splicing factor 3b, subunit 1, 155kD, SF3B155, SF3b155PRP10, spliceosome associated protein 155, Spliceosome-associated protein 155, splicing factor 3B subunit 1, splicing factor 3b, subunit 1, 155kDa | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 23451 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SF3B1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LDSTGYYDQEIYGGSDSRFAGYVTSIAATELEDDDDDYSSSTSLLGQKKPGYHAPVALLNDIPQSTEQYDPFAEHRPPKIADRED | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction