missing translation for 'onlineSavingsMsg'
Learn More

SAR405 R enantiomer, MedChemExpress

Product Code. 16465198
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16465198

Brand: Medchem Express HY12481A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SAR405 R enantiomer is the less active enantiomer of SAR405. SAR405 is a PIK3C3/Vps34 inhibitor.

SAR405 R enantiomer is the less active enantiomer of SAR405. SAR405 is a PIK3C3/Vps34 inhibitor.

Specifications

Chemical Name or Material SAR405 R enantiomer
Physical Form Solid
CAS 1946010-79-2
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H21ClF3N5O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 100 mg/mL (225.30 mM; Need ultrasonic)
SMILES O=C1C=C(N2[C@H](C)COCC2)N=C3N1CC[C@H](C(F)(F)F)N3CC4=CC(Cl)=CN=C4
Molecular Weight (g/mol) 443.85
Formula Weight 443.85
Percent Purity 99.78%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.