missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Furin Monoclonal Antibody (222728)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA560992

Product Code. 30534320

  • 589.00 € / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Furin is a membrane-associated, calcium-dependent, serine protease that belongs to the subtilisin-like prohormone convertase (PC) family. Members of this family of cellular enzymes cleave most prohormones and neuropeptide precursors. Numerous other cellular proteins, some viral proteins, and bacterial toxins that are transported by the constitutive secretory pathway are also targeted for maturation by PCs. Furin and other PC family members share structural similarities which include a heterogeneous ~10 kDa amino-terminal proregion, a highly conserved ~55 kDa subtilisin-like catalytic domain, and carboxyl-terminal domain that is heterogeneous in length and sequence. These enzymes become catalytically active following proregion cleavage within the appropriate cellular compartment. Furin is the only known PC to possess a transmembrane domain. Cleavage of target proteins occurs at the carboxyl-terminus of the furin consensus sequence, RX(K/R)R. It has been shown that the acidic peptide sequence, C771PSDSEEDEG780, localizes furin to the trans-Golgi-network. Phosphorylation of serine residues within this region modulates intracellular routing of furin protein. An additional signaling domain includes the tetrapeptide sequence, Y759KGL762, which directs internalization from the cell surface.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Furin
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P09958
FURIN
Mouse myeloma cell line NS0-derived recombinant human Furin, Asp108-Glu715.
500 μg
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin)
222728
Unconjugated
FURIN
9130404I01Rik; dibasic processing enzyme; dibasic-processing enzyme; FES upstream region; FUR; Furin; furin (paired basic amino acid cleaving enzyme); furin, membrane associated receptor protein; furin, paired basic amino acid cleaving enzyme; furin; LOW QUALITY PROTEIN: furin; Pace; Paired basic amino acid cleaving enzyme (furin); paired basic amino acid residue cleaving enzyme; paired basic amino acid residue-cleaving enzyme; Pcsk3; prohormone convertase 3; proprotein convertase subtilisin/kexin type 3; Proprotein convertase subtilisin/kexin type 3 (paired basic amino acid cleaving enzyme, furin, membrane associated receptor protein); proprotein convertase subtilisin/kexin type3; SPC1; subtilisin pro-protein processing enzyme
Mouse
Protein A/G
RUO
5045
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.