missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-COV-2 NSP13 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00 € - 349.00 €
Specifications
| Antigen | SARS-COV-2 NSP13 |
|---|---|
| Concentration | 400 μg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234967
|
Proteintech
29467-1-AP-150UL |
150 μL |
410.00 € 349.00 € / 150µL Save 61.00 € 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236660
|
Proteintech
29467-1-AP-20UL |
20 μL |
145.00 € 129.00 € / 20µL Save 16.00 € 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SARS-COV-2 NSP13 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of sars-cov-2 SARS-COV-2 NSP13. This antibody recognizes virus antigen. The SARS-COV-2 NSP13 antibody has been validated for the following applications: ELISA analysis.Specifications
| SARS-COV-2 NSP13 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 400 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 43740578 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title