missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Nucleocapsid Recombinant Rabbit Monoclonal Antibody (84C4A), Biotin, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537208
This item is not returnable.
View return policy
Description
SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.
Specifications
| SARS-CoV-2 Nucleocapsid | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.02% sodium azide | |
| P0DTC9 | |
| N | |
| SARS-CoV-2 nucleoprotein (NP) specific peptide. | |
| 200 μL | |
| Primary | |
| Virus | |
| Antibody | |
| IgG1 κ |
| ELISA | |
| 84C4A | |
| Biotin | |
| N | |
| COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N | |
| Rabbit | |
| Protein A | |
| RUO | |
| 43740575 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction