missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Spike Protein Monoclonal Antibody (OTI3A2), TrueMAB™, OriGene

Mouse Monoclonal Antibody

242.00 € - 540.87 €

Specifications

Antigen SARS-CoV-2 Spike Protein
Clone OTI3A2
Concentration 1 mg/mL
Applications ELISA, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17519224
Encompass_Preferred
View Documents
Origene Technologies
TA814466S
30 μL
242.00 €
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17599214
Encompass_Preferred
View Documents
Origene Technologies
TA814466
100 μL
540.87 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.

SARS-CoV-2 Spike Protein Monoclonal antibody specifically detects SARS-CoV-2 Spike Protein in Virus samples. It is validated for ELISA, Western Blot
Specifications

Specifications

SARS-CoV-2 Spike Protein
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Recombinant SARS-CoV-2 Spike-RBD protein produced in E.coli.
Primary
-20° C, Avoid Freeze/Thaw Cycles
OTI3A2
ELISA, Western Blot
Unconjugated
Mouse
Virus
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
IgG2b
Protein A/G
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.