Learn More
Invitrogen™ SARS-CoV-2 Spike Protein RBD Omicron (BA.4/BA.5/BA.2.75) Chimeric Recombinant Human Monoclonal Antibody (D003)

Human Recombinant Monoclonal Antibody
279.00 € - 569.00 €
Specifications
| Antigen | SARS-CoV-2 Spike Protein RBD Omicron (BA.4/BA.5/BA.2.75) Chimeric |
|---|---|
| Clone | D003 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30535673
![]() |
Invitrogen™
MA560600 |
100 μg |
569.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30534033
|
Invitrogen™
MA560601 |
20 μg |
279.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifically recognize the SARS-CoV-2 Omicron BA.4/BA.5/BA.5.2 Spike RBD and ECD (Can block the binding between RBD and ACE2). No cross-reactivity with SARS-CoV-2 wild type, Alpha, Beta, Delta variant. Has Medium cross-reactivity in ELISA with SARS-CoV-2 Omicron BA.2.75 Spike RBD.. Has cross-reactivity in ELISA with: SARS-CoV-2 Omicron (BQ.1.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike RBD Protein; SARS-CoV-2 Omicron (BF.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4.6/BF.7) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.4.6) Spike S1+S2 trimer Protein; No cross-reactivity in ELISA with: SARS-CoV-2 Omicron (XBB.1.16) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike RBD Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.3.20) Spike RBD Protein; SARS-CoV-2 Omicron (B.1.1.529) S1+S2 trimer Protein; SARS-CoV-2 Omicron (B.1.1.529) Spike RBD Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike RBD Protein; SARS-CoV-2 Beta (B.1.351)...
The severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) variant Omicron (B 1 1 529) has attracted great concerns since its identification in South Africa Omicron is the fifth variant of concern (VOC) after Alpha (B 1 1 7), Beta (B 1 351), Gamma (P 1) and Delta (B 1 617 2) Omicron variant harbors >30 substitutions/deletions/insertions in the spike, including 15 substitutions in receptor-binding domain (RBD) [PMID: 35397623].Specifications
| SARS-CoV-2 Spike Protein RBD Omicron (BA.4/BA.5/BA.2.75) Chimeric | |
| 1 mg/mL | |
| ELISA, Neutralization | |
| Unconjugated | |
| Human | |
| RUO | |
| PBS with no preservative | |
| Recombinant SARS-CoV-2 Omicron BA.4/BA.5 Spike ECD Protein. | |
| Antibody |
| D003 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Virus | |
| 2019-nCoV Spike; COVID-19 Spike; SARS-CoV-2 Spike | |
| Primary | |
| Protein A |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.