missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike RBD LlaMABodyTM Bivalent VHH HuIgG2 Fusion Antibody, R&D Systems™

Llama Monoclonal Antibody
178.00 € - 468.00 €
Specifications
| Antigen | Spike RBD |
|---|---|
| Clone | L009.2.69H |
| Dilution | Blockade of Receptor-ligand Interaction |
| Applications | Blocking Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18782377
![]() |
R&D Systems
LMAB10870-SP |
25 μg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772607
![]() |
R&D Systems
LMAB10870-100 |
100 μg |
468.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Blocking AssaySpecifications
| Spike RBD | |
| Blockade of Receptor-ligand Interaction | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Human embryonic kidney cell HEK293-derived SARS-CoV-2 Spike RBD as immunogen for bivalent Llama VHH-Human IgG2 Fusion Antibody., Arg319-Phe541, Accession # YP_009724390.1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| L009.2.69H | |
| Blocking Assay | |
| Unconjugated | |
| Llama | |
| SARS-CoV-2 | |
| YP_009724390.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title