missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike S2 Subunit Antibody, R&D Systems™

Rabbit Monoclonal Antibody
215.00 € - 488.00 €
Specifications
| Antigen | SARS-CoV-2 Spike S2 |
|---|---|
| Clone | 2824A |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724277
![]() |
R&D Systems
MAB10889-SP |
25 μg |
215.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18743297
![]() |
R&D Systems
MAB10889-100 |
100 μg |
488.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SARS-CoV-2 Spike S2 Monoclonal antibody specifically detects SARS-CoV-2 Spike S2 in SARS-CoV-2 samples. It is validated for Western Blot, Immunohistochemistry, ImmunocytochemistrySpecifications
| SARS-CoV-2 Spike S2 | |
| Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| COVID-19, S2 protein, SARS-CoV-2, SARS-CoV-2 Spike, Spike glycoprotein Subunit 2, spike protein, Spike protein S2, structural protein, surface glycoprotein | |
| Chinese Hamster Ovary cell line CHO-derived human SARS-CoV-2 Spike 2 protein, Met1-Lys1211, Accession # YP_009724390.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2824A | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| SARS-CoV-2 | |
| YP_009724390.1 | |
| 43740568 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title