missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS Spike Protein Antibody (16F1071R) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
262.00 € - 536.00 €
Specifications
| Antigen | SARS Spike Protein |
|---|---|
| Clone | 16F1071R |
| Concentration | 1.0 mg/mL |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636521
|
Novus Biologicals
NBP2-90998 |
0.1 mg |
536.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654839
|
Novus Biologicals
NBP2-90998SS |
0.025 mg |
262.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SARS Spike Protein Monoclonal antibody specifically detects SARS Spike Protein in SARS-CoV samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SARS Spike Protein | |
| 1.0 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| SARS-CoV | |
| E2, E2 glycoprotein, Human coronavirus spike glycoprotein, Peplomer protein, S, S glycoprotein, SARS coronavirus S protein, SARS coronavirus Spike Protein, SARS CoV, SARS CoV S glycoprotein, SARS CoV Spike protein, SARSCoV, SARS-CoV S protein, SARS-COV Spike, Severe acute respiratory syndrome spike glycoprotein, Severe acute respiratory syndrome virus spike glycoprotein, Spike glycoprotein, VGL2 | |
| IgG1 κ | |
| Immunogen affinity purified |
| 16F1071R | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Immunology, Virology Bacteria and Parasites | |
| PBS | |
| This recombinant antibody was developed by immunizing mice with a synthetic peptide corresponding to amino acids 19-35 (CTTFDDVQAPNYTQHTS) from the S (Spike glycoprotein)(Spike Protein 1) for the Human SARS coronavirus (Genbank accession no. NP_828851.1) | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title