missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 624.00 €
Specifications
| Antigen | SARS2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18454582
|
Novus Biologicals
NBP2-33666-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18112135
|
Novus Biologicals
NBP2-33666 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SARS2 Polyclonal specifically detects SARS2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SARS2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9NP81 | |
| 54938 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QIYNIDPARFKDLNLAGTAEVGLAGYFMDHTVAFRDLPVRMVCSSTCYRAETNTGQEPRGLYRVHHFTKVEMFGVTGPGL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EC 6.1.1.11, FLJ20450, mitochondrial seryl-tRNA synthetase, mtSerRS, SARS, serine tRNA ligase 2, mitochondrial, Serine--tRNA ligase, serine-tRNA ligase, mitochondrial, SerRS, SerRSmtSARSM, SERS, seryl-tRNA synthetase 2, seryl-tRNA synthetase 2, mitochondrial, seryl-tRNA synthetase, mitochondrial, Seryl-tRNA(Ser/Sec) synthetase, SYS | |
| SARS2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title