Learn More
Sartorius Vivacon™ 2 DNA Concentrator
Shop All Sartorius Products

Description
Vivacon 2 are centrifugal devices for DNA concentration and buffer exchange applications for samples up to 2 ml. For optimal performance with very diluted samples, e.g. forensic specimens, Vivacon 2 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux. High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon 2 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as protein fractionation (peptide removal). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.
- Convenient handling in fixed-angle rotors
- Low binding material
- Reverse spin possibility
- Graduation and MWCO printed on device
- Complete and highly reproducible concentrate recovery
Specifications
Specifications
| Concentration | 50ng/mL |
| Concentration Time | 30 min. |
| Filter Type | Ultrafiltration |
| Material | Polycarbonate |
| Material (Membrane) | Cellulose |
| MWCO | 100 kDa |
| Sample Volume | 2 mL |
| Sterility | Non-sterile |
| Filtration Area | 0.95 cm2 |
| For Use With (Application) | DNA concentration, desalting, buffer exchange, fractionation |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.