missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivacon™ 2 DNA Concentrator
BIOPHARMA

Product Code. 11780846 Shop All Sartorius Products
Click to view available options
MWCO:
10 kDa
100 kDa
2 kDa
30 kDa
50 kDa
Quantity:
100 Pack
25 Pack
500 Pack
Product Line:
Hydrosart
Vivacon 2
Unit Size:
Pack of 100
Pack of 25
Pack of 500
This item is not returnable. View return policy

Product Code. 11780846

Brand: Sartorius VN02H43

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Centrifugal units for fast DNA concentration and buffer exchange of 2 ml samples

Vivacon 2 are centrifugal devices for DNA concentration and buffer exchange applications for samples up to 2 ml. For optimal performance with very diluted samples, e.g. forensic specimens, Vivacon 2 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux. High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon 2 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as protein fractionation (peptide removal). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.

  • Convenient handling in fixed-angle rotors
  • Low binding material
  • Reverse spin possibility
  • Graduation and MWCO printed on device
  • Complete and highly reproducible concentrate recovery
TRUSTED_SUSTAINABILITY

Specifications

Concentration 50ng/mL
Concentration Time 30 min.
Filter Type Ultrafiltration
Material Polycarbonate
Material (Membrane) Cellulose
MWCO 100 kDa
Sample Volume 2 mL
Sterility Non-sterile
Filtration Area 0.95 cm2
For Use With (Application) DNA concentration, desalting, buffer exchange, fractionation
Max. Force 2500xG
Product Line Vivacon 2
Quantity 500 Pack
Sample Type DNA, peptide
Type DNA Concentrator
Volume (Metric) 2 mL
Volume (Metric) Hold Up 10 μL
Dimensions (L x W) 4.92 x 0.62 in. (12.5 x 1.6cm)
Length (English) 4.92 in.
Length (English) Back Spin 4.52 in.
Length (Metric) 12.5 cm
Length (Metric) Back Spin 11.5 cm
Width (English) 0.62 in.
Width (Metric) 1.6 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.