missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaflow™ 50R Crossflow Cassettes
BIOPHARMA

Product Code. 15983030 Shop All Sartorius Products
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
30 kDa
5 kDa
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Unit Size
15983030 100 kDa Each
15730258 5 kDa Each
15198279 30 kDa Each
15178279 10 kDa Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15983030 Supplier Sartorius Supplier No. VF05H4
  • ONLINE EXCLUSIVE PRICE
    335.00 €
    313.00 € / Pack of 1
    Save 22.00 €
    6% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings

Unique, Plug and Play laboratory crossflow devices for concentrating samples like viruses and antibodies

  • Reusable device with low-binding Hydrosart™ (modified regenerated cellulose) membrane
  • For concentrating up to 1L samples
  • Active membrane area per device is 50cm2
  • One unit comes with all necessary accessories for running device with laboratory pump and size 16 pump head
  • For speeding up concentration, two cassettes can be run simultaniously
  • Acrylic main housing and flow channel
  • Polypropylene membrane support
  • Nylon fittings and PVC tubing
  • Compatible with 1M NaOH for cleaning and depyrogenation
  • High flow rate and recovery
  • L x W x H: 100x 24 x 100mm
  • Pump flow rate: 200 to 400mL/min
  • Maximum pressure: 45psi
  • Maximum temperature: 60°C
  • Antibody/recombinant protein concentration/diafiltration in biopharma research
  • Concentration of small production lots of proteins for diagnostic purposes
  • Concentration of viruses from cell culture supernatants or of environmental samples
  • Nanoparticle concentration
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Format Cassette
Material Acrylic
Material (Membrane) Cellulose
MWCO 100 kDa
Sterility Non-sterile
For Use With (Application) Concentrating viruses and antibodies
Includes Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings
Sample Type Protein, Virus, Nanoparticles
Type Crossflow Cassette
Volume (Metric) 1 L
For Use With (Equipment) Labscale TFF System
Sample Volume 1 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.