Learn More
SATB2 Antibody (CL0321) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52915AF488
Product Code. 30292264
  • 737.00 € / 0.10mL
Description
SATB2 Monoclonal antibody specifically detects SATB2 in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SATB2
Monoclonal
Alexa Fluor 488
DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2
This antibody was developed using a recombinant protein derived from Q9UPW6, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
23314
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0321
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Rat
Purified