missing translation for 'onlineSavingsMsg'
Learn More

SB 202190, MedChemExpress

Product Code. 16454826
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454826 100 mg 100mg
16470966 200 mg 200mg
16420147 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16454826 Supplier Medchem Express Supplier No. HY10295
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SB 202190 is a selective p38 MAP kinase inhibitor with IC50s of 50 nM and 100 nM for p38α and p38β2, respectively. SB 202190 binds to the ATP pocket of the active recombinant human p38 kinase with a Kd of 38 nM. SB 202190 has anti-cancer activity and rescued memory deficits. SB202190 induces autophagy.

SB 202190 is a selective p38 MAP kinase inhibitor with IC50s of 50 nM and 100 nM for p38α and p38β2, respectively. SB 202190 binds to the ATP pocket of the active recombinant human p38 kinase with a Kd of 38 nM. SB 202190 has anti-cancer activity and rescued memory deficits. SB202190 induces autophagy.

Specifications

Chemical Name or Material SB 202190
CAS 152121-30-7
Color Light Nude
Purity Grade Notes Research
Molecular Formula C20H14FN3O
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (301.80 mM; Need ultrasonic)
SMILES FC1=CC=C(C2=C(C3=CC=NC=C3)NC(C4=CC=C(O)C=C4)=N2)C=C1
Molecular Weight (g/mol) 331.34
Formula Weight 331.34
Percent Purity 98.56%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.