missing translation for 'onlineSavingsMsg'
Learn More

SB 228357, Tocris Bioscience™

Product Code. 18095282
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095282 10 mg 10mg
18094092 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095282 Supplier Tocris Bioscience Supplier No. 1375/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5-HT2C/2B antagonist/inverse agonist

5-HT2C/2B receptor antagonist (pKi values are 7.0, 8.1 and 9.1 at 5-HT2A, 2B and 2C receptors respectively). Displays inverse agonism in a 5-HT-stimulated PI hydrolysis model of 5-HT2C receptor function. Orally active in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181629-93-6
Molecular Formula C22H17F4N3O2
Molecular Weight (g/mol) 431.391
InChI Key RRJLJKRFFRZRAF-UHFFFAOYSA-N
Synonym unii-5rdn2e8e97, tocris-1375, chembl14276, n-3-fluoro-5-3-pyrindyl phenyl-2,3-dihydro-5-methoxy-6-trifluoromethyl-1h-indole-1-carboxamide, n-3-fluoro-5-pyridin-3-ylphenyl-5-methoxy-6-trifluoromethyl-2,3-dihydroindole-1-carboxamide, n-3-fluoro-5-3-pyridinyl phenyl-2,3-dihydro-5-methoxy-6-trifluoromethyl-1h-indole-1-carboxamide, lopac-s-2318, d05nwh, gtpl192, n-3-3-pyridinyl-5-fluorophenyl-5-methoxy-6-trifluoromethyl indoline-1-carboxamide
PubChem CID 443390
ChEBI CHEBI:8979
IUPAC Name N-(3-fluoro-5-pyridin-3-ylphenyl)-5-methoxy-6-(trifluoromethyl)-2,3-dihydroindole-1-carboxamide
SMILES COC1=C(C=C2C(=C1)CCN2C(=O)NC3=CC(=CC(=C3)C4=CN=CC=C4)F)C(F)(F)F

Specifications

Quantity 10 mg
Formula Weight 431.39
Percent Purity >99%
Chemical Name or Material SB 228357
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.