missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SBSN Antibody (SBSN/7961), Novus Biologicals™
Mouse Monoclonal Antibody
232.00 € - 462.00 €
Specifications
| Antigen | SBSN |
|---|---|
| Clone | SBSN/7961 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662304
|
Novus Biologicals
NBP3-23822-20ug |
20 μg |
232.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654015
|
Novus Biologicals
NBP3-23822-100ug |
100 μg |
462.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SBSN Monoclonal antibody specifically detects SBSN in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| SBSN | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| HLAR698, MGC75533, suprabasin, UNQ698 | |
| Recombinant fragment of human SBSN protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| SBSN/7961 | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 374897 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title