missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SC 236
GREENER_CHOICE

Product Code. 18045112
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18045112

Brand: Tocris Bioscience™ 3919/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective cyclooxygenase-2 (COX-2) inhibitor

Selective COX-2 inhibitor (IC50 values are 0.005 and 17.8 μM for COX-2 and COX-1 respectively). Displays anti-inflammatory properties and potent antimetastatic activity against both experimental metastases and spontaneous metastases arising following primary tumor excision.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170569-86-5
Molecular Formula C16H11ClF3N3O2S

Specifications

Quantity 10 mg
Formula Weight 401.79
Purity >99%
Molecular Weight (g/mol) 401.788
Chemical Name or Material SC 236
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.