missing translation for 'onlineSavingsMsg'
Learn More

Scandium Standard For ICP, 1000μg/mL, SPEX CertiPrep™

Product Code. 10336222 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Product Code. Quantity unitSize
10336222 250 mL 250mL
1 options
This item is not returnable. View return policy

Product Code. 10336222

Brand: SPEX Certiprep PLSC22T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sc1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 12060-08-1, 7697-37-2, 7732-18-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 2.0, 97.7, 0.3
Analyte or Component Names Sc
Molecular Formula O3Sc2
MDL Number MFCD00011223
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key HJGMWXTVGKLUAQ-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [O--].[O--].[O--].[Sc+3].[Sc+3]
Element Sc
IUPAC Name discandium(3+) trioxidandiide
Solvent or Matrix 2% HNO3
Molecular Weight (g/mol) 137.91
Quantity 250 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) 1000mg/L
Chemical Name or Material Scandium Standard, For AA and ICP
Show More Show Less
compliance-icons
Product Identifier
  • 1000 µg/mL Scandium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.