missing translation for 'onlineSavingsMsg'
Learn More

Scandium, plasma standard solution, Specpure™ Sc 1000μg/mL

Product Code. 11338667
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338667 50 mL 50mL
11328667 100 mL 100mL
11455070 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338667 Supplier Thermo Scientific Alfa Aesar Supplier No. 035769.AD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Sc 1000μg/mL | CAS: 7440-20-2 | Sc | 44.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-20-2
Molecular Formula Sc
Molecular Weight (g/mol) 44.96
MDL Number MFCD00016323
InChI Key SIXSYDAISGFNSX-UHFFFAOYSA-N
PubChem CID 23952
ChEBI CHEBI:33330
IUPAC Name scandium
SMILES [Sc]

Specifications

Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Quantity 50 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 5%, Scandium(III) oxide: 0.15%, Water: 94.85%
Chemical Name or Material Scandium, plasma standard solution

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.