missing translation for 'onlineSavingsMsg'
Learn More

Scandium Std - Sc @ 5000μg/g in 75 cSt Oil

Product Code. 30323884
Change view
Click to view available options
Quantity:
200 g
400 g
Unit Size:
200g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323884 200 g 200g
30314911 400 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30323884 Supplier VHG Labs Supplier No. VHGOSC5000200G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Scandium
CAS 7440-20-2
Analyte or Component Names Scandium
Shipping Condition Room Temperature
Molecular Formula Sc
Concentration 5000 μg/g
Synonym Sc
InChI Formula InChI=1 S/Sc
Solution Type Single Solution
SMILES [Sc]
Element Sc
IUPAC Name scandium
Solvent or Matrix 75 cSt Oil
Industry Type Industrial QC
Molecular Weight (g/mol) 44.96
Quantity 200 g
Formula Weight 44.956
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.