missing translation for 'onlineSavingsMsg'
Learn More

SCFD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16850344 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16850344 20 μL 20µL
16840344 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16850344 Supplier Proteintech Supplier No. 125691AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The de novo synthesis of pyrimidine nucleotides is required for mammalian cells to proliferate. This gene encodes a trifunctional protein which is associated with the enzymatic activities of the first 3 enzymes in the 6-step pathway of pyrimidine biosynthesis: carbamoylphosphate synthetase (CPS II), aspartate transcarbamoylase, and dihydroorotase. This protein is regulated by the mitogen-activated protein kinase (MAPK) cascade, which indicates a direct link between activation of the MAPK cascade and de novo biosynthesis of pyrimidine nucleotides.

Specifications

Antigen SCFD1
Applications Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SCFD1
Gene Accession No. Q62991, Q8BRF7, Q8WVM8
Gene Alias C14orf163, KIAA0917, RA410, SCFD1, SLY1, SLY1 homolog, Sly1p, STXBP1L2
Gene Symbols Scfd1
Host Species Rabbit
Immunogen SCFD1 Fusion Protein Ag3234
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23256, 54350, 76983
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.