missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The de novo synthesis of pyrimidine nucleotides is required for mammalian cells to proliferate. This gene encodes a trifunctional protein which is associated with the enzymatic activities of the first 3 enzymes in the 6-step pathway of pyrimidine biosynthesis: carbamoylphosphate synthetase (CPS II), aspartate transcarbamoylase, and dihydroorotase. This protein is regulated by the mitogen-activated protein kinase (MAPK) cascade, which indicates a direct link between activation of the MAPK cascade and de novo biosynthesis of pyrimidine nucleotides.
Specifications
Specifications
| Antigen | SCFD1 |
| Applications | Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.37 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | SCFD1 |
| Gene Accession No. | Q62991, Q8BRF7, Q8WVM8 |
| Gene Alias | C14orf163, KIAA0917, RA410, SCFD1, SLY1, SLY1 homolog, Sly1p, STXBP1L2 |
| Gene Symbols | Scfd1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?