missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SCH 39166 hydrobromide
GREENER_CHOICE

Product Code. 30001367
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001367 10 mg 10mg
18004112 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001367 Supplier Tocris Bioscience™ Supplier No. 2299/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity D1-like antagonist

High affinity dopamine D1/D5 receptor antagonist; displays Ki values of 1.2, 2, 980, 5520, 80 and 731 nM for binding to D1, D5, D2, D4, 5-HT and α2a receptors, respectively.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1227675-51-5
Molecular Formula C19H21BrClNO

Specifications

Quantity 10 mg
Formula Weight 394.73
Purity >99%
Molecular Weight (g/mol) 394.737
Chemical Name or Material SCH 39166 hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.