missing translation for 'onlineSavingsMsg'
Learn More

Schisandrin A, MedChemExpress

Product Code. 16459068
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459068 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16459068 Supplier Medchem Express Supplier No. HYN0693
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Schisandrin A inhibits CYP3A activity with an IC50 of 6.60 μM and Ki of 5.83 μM, respectively.

Schisandrin A inhibits CYP3A activity with an IC50 of 6.60 μM and Ki of 5.83 μM, respectively.

Specifications

Chemical Name or Material Schisandrin A
CAS 61281-38-7
Color White
Purity Grade Notes Research
Molecular Formula C24H32O6
Quantity 10 mg
Synonym Schizandrin-A Wuweizisu-A Deoxyschizandrin
Solubility Information DMSO : 50 mg/mL (120.05 mM; Need ultrasonic)
SMILES C[C@H]1CC2=CC(OC)=C(OC)C(OC)=C2C3=C(OC)C(OC)=C(OC)C=C3C[C@H]1C
Molecular Weight (g/mol) 416.51
Formula Weight 416.51
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.