missing translation for 'onlineSavingsMsg'
Learn More

Porvair Sciences™ Science Polystyrene Black Microplate

Product Code. 12394108
Encompass_Preferred
Change view
Click to view available options
Surface Treatment:
Non-treated
Tissue Culture
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Treatment unitSize
12394108 Tissue Culture Pack of 100
12449337 Non-treated Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12394108 Supplier Porvair Sciences™ Supplier No. 223009

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Square wells maximise liquid volume per well (120μL/well)
  • White plate prevents cross talk in light emitting assays
  • Black plate gives low autofluorescence background
  • Tissue culture treated plates for anchorage dependent cells
  • Total well volume 120μL
  • Dimensions [w x d x h] 3.75mm x 3.75mm x 11.4mm (well), 85.5mm x 127.5mm x 14.8mm (plate)
  • 24 x 16 well array

Specifications

Format Standard
Array 16 x 24
Color Black
Product Type Microplate
Material Polystyrene
No. of Wells 384
Surface Treatment Tissue Culture
Total Volume (Metric) Well 120 μL
For Use With (Application) Tissue Culture Treated
Includes Lid
No. per Pack 100
Quantity 100 Each
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.