missing translation for 'onlineSavingsMsg'
Learn More

Scopolamine Sulfate, TRC

Product Code. 30412435
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30412435 250 mg
30418333 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30412435 Supplier Toronto Research Chemicals Supplier No. TRCS200040250MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866926-81-0
Molecular Formula C17H21NO7S
Molecular Weight (g/mol) 383.42
Synonym α-[(Sulfooxy)methyl]benzeneacetic Acid C-[(1α,2α,4α,5α,7β)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl] Ester
SMILES CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](COS(=O)(=O)O)c4ccccc4

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H21NO7S/c1-18-13-7-11(8-14(18)16-15(13)25-16)24-17(19)12(9-23-26(20,21)22)10-5-3-2-4-6-10/h2-6,11-16H,7-9H2,1H3,(H,20,21,22)/t11-,12-,13-,14+,15-,16+/m1/s1
Formula Weight 383.1039
Chemical Name or Material Scopolamine Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.