Learn More
Invitrogen™ SCP2 Recombinant Rabbit Monoclonal Antibody (24GB790)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557270
 View more versions of this product
Product Code. 30274988
  • 146.00 € / 20µL
Description
SCP2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes two proteins: sterol carrier protein X (SCPx) and sterol carrier protein 2 (SCP2), as a result of transcription initiation from 2 independently regulated promoters. The transcript initiated from the proximal promoter encodes the longer SCPx protein, and the transcript initiated from the distal promoter encodes the shorter SCP2 protein, with the 2 proteins sharing a common C-terminus. Evidence suggests that the SCPx protein is a peroxisome-associated thiolase that is involved in the oxidation of branched chain fatty acids, while the SCP2 protein is thought to be an intracellular lipid transfer protein. This gene is highly expressed in organs involved in lipid metabolism, and may play a role in Zellweger syndrome, in which cells are deficient in peroxisomes and have impaired bile acid synthesis. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SCP2
Recombinant Monoclonal
Unconjugated
SCP2
AA409774; AA409893; C76618; C79031; NLTP; nonspecific lipid transfer protein; Non-specific lipid-transfer protein; NSLIPTR; ns-LTP; NSL-TP; propanoyl-CoA C-acyltransferase; SCP2; SCP-2; SCP-CHI; SCPX; SCP-X; Sterol carrier protein 2; sterol carrier protein 2, liver; sterol carrier protein X
Rabbit
Affinity chromatography
RUO
20280, 25541, 6342
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB790
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P11915, P22307, P32020
SCP2
A synthesized peptide derived from human Sterol carrier protein 2 (aa 14-64).
20 μL
Primary
Human, Mouse, Rat