missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP2/SYCP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94045-0.1ml
This item is not returnable.
View return policy
Description
SCP2/SYCP2 Polyclonal antibody specifically detects SCP2/SYCP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SCP2/SYCP2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| NLTP, non-specific lipid-transfer protein, NSL-TP, Propanoyl-CoA C-acyltransferase, SCP-2, SCP-CHI, SCPX, SCP-X, Sterol Carrier Protein 2, Sterol carrier protein X | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1231-1530 of human SYCP2 (NP_055073.2). EIESPHINENYIQSKREESHLASSLSKSSEGREKTWFDMPCDATHVSGPTQHLSRKRIYIEDNLSNSNEVEMEEKGERRANLLPKKLCKIEDADHHIHKMSESVSSLSTNDFSIPWETWQNEFAGIEMTYETYERLNSEFKRRNNIRHKMLSYFTTQSWKTAQQHLRTMNHQSQDSRIKKLDKFQFIIIEELENFEKDSQSLKDLEKEFVDFWEKIFQKFSAYQKSEQQRLHLLKTSLAKSVFCNTDSEETVFTSEMCLMKEDMKVLQDRLLKDMLEEELLNVRRELMSVFMSHERNANV | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cell Cycle and Replication | |
| 10388 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction