missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDCBP Mouse anti-Human, Clone: UMAB69, liquid, UltraMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies UM500057
This item is not returnable.
View return policy
Description
The protein encoded by this gene was initially identified as a molecule linking syndecan-mediated signaling to the cytoskeleton. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. This protein may also affect cytoskeletal-membrane organization, cell adhesion, protein trafficking, and the activation of transcription factors. The protein is primarily localized to membrane-associated adherens junctions and focal adhesions but is also found at the endoplasmic reticulum and nucleus. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| SDCBP | |
| Monoclonal | |
| Unconjugated | |
| SDCBP | |
| MDA-9, MDA9, ST1, SYCL, TACIP18 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6386 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| UMAB69 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00560 | |
| SDCBP | |
| Full length human recombit protein of human SDCBP produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction