missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDH Assembly Factor 1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00 € - 532.00 €
Specifications
| Antigen | SDH Assembly Factor 1 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18659276
|
Novus Biologicals
NBP2-41176-0.025mg |
0.025 mg |
189.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18118917
|
Novus Biologicals
NBP2-41176 |
0.1 mg |
532.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SDH Assembly Factor 1 Polyclonal specifically detects SDH Assembly Factor 1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| SDH Assembly Factor 1 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 644096 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SDHAF1. The immunogen is located within the last 50 amino acids of SDHAF1. Amino Acid Squence: DSPRNPHDSTGAPETRPD | |
| Primary | |
| SDHAF1 antibody is predicted to not cross-react with other SDHAF protein family members. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| LYR Motif Containing 8, LYR Motif-Containing Protein 8, LYRM8, SDHAF1, Succinate Dehydrogenase Assembly Factor 1, Mitochondrial, Succinate Dehydrogenase Complex Assembly Factor 1 | |
| SDHAF1 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title