missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHAF1 Monoclonal Antibody (OTI1E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809691
This item is not returnable.
View return policy
Description
SDHAF1 (Succinate dehydrogenase complex assembly factor 1) is one of the subunits of the succinate dehydrogenase (SDH) complex (or complex II) of the mitochondrial respiratory chain. SDHAF1 resides in the mitochondria, and is essential for SDH assembly, but does not physically associate with the complex in vivo. Mutations in this gene are associated with SDH-defective infantile leukoencephalopathy (mitochondrial complex II deficiency). Unlike the related protein SDHAF2, SDHAF1 is not thought to be a tumor suppressor.Specifications
| SDHAF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| A6NFY7 | |
| SDHAF1 | |
| Full length human recombit protein of human SDHAF1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1E6 | |
| Unconjugated | |
| SDHAF1 | |
| LYRM8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 644096 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction