Learn More
SDHB Antibody (CL0347) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52920CL1
Product Code. 30297583
  • 1175.00 € / 0.10mL
Description
SDHB Monoclonal antibody specifically detects SDHB in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SDHB
Monoclonal
CoraFluor 1
EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip)
This antibody was developed using a recombinant protein derived from P21912, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
6390
Store at 4⪚C in the dark. Do not freeze.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0347
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified