missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99066-50ul
This item is not returnable.
View return policy
Description
SDHB Polyclonal antibody specifically detects SDHB in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SDHB | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip) | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SDHB. | |
| 50 μg | |
| Cancer | |
| 6390 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction