Learn More
Invitrogen™ SDHC Recombinant Rabbit Monoclonal Antibody (24GB1075)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557707
 View more versions of this product
Product Code. 30275931
  • 198.00 € / 20µL
Description
SDHC Recombinant Monoclonal Antibody for Western Blot, ICC/IF
This gene encodes one of four nuclear-encoded subunits that comprise succinate dehydrogenase, also known as mitochondrial complex II, a key enzyme complex of the tricarboxylic acid cycle and aerobic respiratory chains of mitochondria. The encoded protein is one of two integral membrane proteins that anchor other subunits of the complex, which form the catalytic core, to the inner mitochondrial membrane. There are several related pseudogenes for this gene on different chromosomes. Mutations in this gene have been associated with paragangliomas. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SDHC
Recombinant Monoclonal
Unconjugated
SDHC
0610010E03Rik; AI316496; AU019277; CYB560; CYBL; cytochrome B large subunit of complex II; integral membrane protein CII-3; integral membrane protein CII-3b; large subunit of cytochrome b; PGL3; QPs1; QPs-1; SDH3; SDHC; succinate dehydrgenase cytochrome b; succinate dehydrogenase 3, integral membrane subunit; succinate dehydrogenase complex subunit C; succinate dehydrogenase complex subunit C integral membrane protein 15kDa; succinate dehydrogenase complex, subunit C, integral membrane protein; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kD; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kDa; succinate dehydrogenase cytochrome b560 subunit, mitochondrial; Succinate-ubiquinone oxidoreductase cytochrome B large subunit
Rabbit
Affinity chromatography
RUO
289217, 6391, 66052
-20°C
Liquid
Western Blot, Immunocytochemistry
24GB1075
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q99643, Q9CZB0
SDHC
A synthesized peptide derived from human SDHC (aa 10-69).
20 μL
Primary
Human, Mouse, Rat