missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl methyl sulfide, 98%, Thermo Scientific™

Product Code. 11415047
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415047 5 g 5g
11425047 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415047 Supplier Thermo Scientific Alfa Aesar Supplier No. B21942.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10359-64-5
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.211
MDL Number MFCD00015193
InChI Key IJRCRFQMYAJPPO-UHFFFAOYSA-N
Synonym 2-methylthio butane, sec-butyl methyl sulfide, butane, 2-methylthio, sulfide, sec-butyl methyl, methyl sec-butyl sulfide, acmc-20ajqw, methyl-sec-butylsulfid, 3-methyl-2-thiapentane, sec-butyl methyl sulfane, 2-methylsulfanyl butane
PubChem CID 93062
IUPAC Name 2-methylsulfanylbutane
SMILES CCC(C)SC

Specifications

Density 0.824
Boiling Point 116°C to 117°C
Flash Point 5°C (41°F)
Odor Unpleasant
Refractive Index 1.447
Quantity 5 g
UN Number UN1993
Beilstein 1730946
Formula Weight 104.22
Percent Purity 98%
Chemical Name or Material sec-Butyl methyl sulfide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.